Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp609 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172536.3 | NP_766124.2 | 5 | 1755 | ||
GenBank mRNA | AK036715.1 | - | 4 | 1488 | ||
BC016271.1 | - | 5 | 1755 | |||
BC058943.1 | - | 4 | 1660 | |||
BC150683.1 | - | 4 | 1464 | 9087594 |