Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172554.2 | NP_766142.2 | 5 | 447 | ||
GenBank mRNA | AK038670.1 | - | 5 | 391 | ||
AK038681.1 | - | 5 | 391 | |||
AK039575.1 | - | 5 | 448 | |||
AK046891.1 | - | 5 | 389 | |||
AK122406.1 | - | 5 | 521 | |||
AK134967.1 | - | 5 | 396 | |||
AK162922.1 | - | 5 | 385 | |||
BC027101.1 | - | 5 | 376 | |||
BC058772.1 | - | 5 | 388 | |||
BC080664.1 | - | 5 | 386 | |||
BC117788.1 | - | 5 | 389 | 40092137 | ||
BC117789.2 | - | 5 | 389 | 40092140 |