Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sipa1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167983.1 | NP_001161455.1 | 5 | 886 | ||
NM_172579.3 | NP_766167.2 | 2 | 690 | |||
GenBank mRNA | AK029889.1 | - | 3 | 716 | ||
AK147591.1 | - | 4 | 1413 | |||
AK171876.1 | - | 5 | 1060 | |||
BC058681.1 | - | 5 | 876 | 6834753 | ||
BC059219.1 | - | 1 | 402 | |||
BC095958.1 | - | 5 | 875 | 5343588 |