Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp697 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172863.4 | NP_766451.3 | 3 | 2205 | ||
GenBank mRNA | AK034880.1 | - | 3 | 2206 | ||
BC092534.1 | - | 3 | 2138 | 30249621 |