Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174874.3 | NP_777363.1 | 3 | 137 | ||
GenBank mRNA | AJ504653.1 | - | 2 | 129 | ||
AK010949.1 | - | 3 | 265 | |||
AK028712.1 | - | 3 | 148 | |||
AK031570.1 | - | 3 | 138 | |||
AK075796.1 | - | 3 | 148 | |||
AK088811.1 | - | 3 | 210 | |||
AK129242.1 | - | 2 | 125 | |||
AK144239.1 | - | 3 | 143 | |||
BC027184.1 | - | 3 | 163 | 4237950 |