Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145967.1 | NP_001139439.1 | 3 | 322 | ||
NM_175029.3 | NP_778194.3 | 3 | 704 | |||
GenBank mRNA | AK134838.1 | - | 3 | 471 | ||
AK156463.1 | - | 3 | 474 | |||
AK170747.1 | - | 3 | 461 | |||
BB851037.1 | - | 3 | 323 | |||
BC058981.1 | - | 3 | 704 | 6827140 |