Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175300.4 | NP_780509.2 | 6 | 1318 | ||
GenBank mRNA | AK033784.1 | - | 6 | 1286 | ||
AK129351.1 | - | 6 | 1271 | |||
AK154465.1 | - | 6 | 1272 | |||
AK170676.1 | - | 6 | 1284 | |||
BC027351.1 | - | 6 | 1266 | |||
BC038267.1 | - | 5 | 630 | |||
BC096381.1 | - | 5 | 1511 | |||
BC137583.1 | - | 6 | 1286 | 8860602 |