Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175389.4 | NP_780598.2 | 2 | 192 | ||
GenBank mRNA | AK089967.1 | - | 2 | 287 | ||
AK145681.1 | - | 2 | 263 | |||
AK146481.1 | - | 2 | 324 | |||
BC022121.1 | - | 1 | 82 | |||
BC090650.1 | - | 2 | 192 | 30651613 | ||
BC098478.1 | - | 2 | 257 | 30033713 | ||
BC141035.1 | - | 2 | 180 | 9055647 |