Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175399.4 | NP_780608.1 | 3 | 1134 | ||
GenBank mRNA | AK083308.1 | - | 3 | 1094 | ||
AK089890.1 | - | 3 | 1135 | |||
AK144062.1 | - | 3 | 1049 | |||
AK167629.1 | - | 3 | 1097 |