Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrgpre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175534.3 | NP_780743.1 | 2 | 1301 | ||
GenBank mRNA | AJ320231.1 | - | 2 | 1012 | ||
AK048528.1 | - | 2 | 1119 | |||
AK133657.1 | - | 2 | 1303 | |||
AK154886.1 | - | 2 | 1096 | |||
BC096766.1 | - | 2 | 1097 | 30102203 | ||
BC138305.1 | - | 2 | 1010 | 8861325 | ||
BC138308.1 | - | 2 | 1010 | 8861328 |
Set Membership: |
G-protein coupled receptor |