Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178707.4 | NP_848822.2 | 3 | 611 | ||
GenBank mRNA | AK042675.1 | - | 3 | 613 | ||
AK129089.1 | - | 3 | 583 | |||
AK143618.1 | - | 3 | 609 | |||
BC094321.1 | - | 1 | 396 | 6401005 | ||
BC118972.1 | - | 2 | 465 | 40098313 | ||
BC118973.1 | - | 2 | 465 | 40098315 |