Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ushbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181418.3 | NP_852083.2 | 10 | 1633 | ||
GenBank mRNA | AK043798.1 | - | 10 | 1571 | ||
AK084049.1 | - | 7 | 1080 | |||
BC026453.1 | - | 10 | 1617 | 4460920 | ||
BC033912.1 | - | 3 | 555 |