Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181549.3 | NP_853527.2 | 11 | 1377 | ||
GenBank mRNA | AK132888.1 | - | 13 | 2245 | ||
AY223868.1 | - | 1585 | ||||
AY223869.1 | - | 1556 | ||||
AY223870.1 | - | 1560 | ||||
AY223871.1 | - | 1466 | ||||
AY223872.1 | - | 1531 | ||||
AY223873.1 | - | 1437 |