Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uggt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198899.2 | NP_942602.2 | 5 | 619 | ||
GenBank mRNA | AK050532.1 | - | 5 | 606 | ||
AK087883.1 | - | 5 | 425 | |||
AK144381.1 | - | 5 | 479 | |||
BC062936.1 | - | 5 | 566 | 6838545 | ||
BC068283.1 | - | 5 | 601 | 30022247 | ||
CK639915.1 | - | 5 | 619 |