Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166350.1 | NP_001159822.1 | 2 | 233 | ||
NM_199314.2 | NP_955018.2 | 2 | 247 | |||
GenBank mRNA | AK149420.1 | - | 2 | 211 | ||
BC024087.1 | - | 2 | 225 | 5101271 | ||
BC096459.1 | - | 2 | 228 | |||
BC096585.1 | - | 2 | 194 | 5097495 | ||
BY730795.1 | - | 2 | 233 |