Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166350.1 | NP_001159822.1 | 3 | 740 | ||
NM_199314.2 | NP_955018.2 | 3 | 754 | |||
GenBank mRNA | AK149420.1 | - | 3 | 718 | ||
BC024087.1 | - | 3 | 732 | 5101271 | ||
BC096459.1 | - | 4 | 646 | |||
BC096585.1 | - | 3 | 701 | 5097495 |