Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gal3st2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199366.4 | NP_955398.3 | 3 | 606 | ||
GenBank mRNA | AK144526.1 | - | 3 | 605 | ||
AK165692.1 | - | 3 | 606 | |||
BC132622.1 | - | 3 | 385 | 40130899 | ||
BC132624.1 | - | 3 | 385 | 40130901 |
Gene Symbol: Gm6086 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039219.2 | NP_001034308.1 | 7 | 1210 | ||
GenBank mRNA | AK145447.1 | - | 7 | 1211 |
Gene Symbol: Gm9994 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205249.1 | NP_001192178.1 | 350 | |||
GenBank mRNA | AY216522.1 | - | 350 |