Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm9994 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205249.1 | NP_001192178.1 | 851 | |||
GenBank mRNA | AY216522.1 | - | 851 |
Gene Symbol: Gal3st2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199366.4 | NP_955398.3 | 4 | 1107 | ||
GenBank mRNA | AK144526.1 | - | 4 | 1106 | ||
AK165692.1 | - | 4 | 1111 | |||
BC132622.1 | - | 4 | 886 | 40130899 | ||
BC132624.1 | - | 4 | 886 | 40130901 |
Gene Symbol: Gm6086 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039219.2 | NP_001034308.1 | 8 | 1711 | ||
GenBank mRNA | AK145447.1 | - | 8 | 1712 |