Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVI5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308248.1 | NP_001295177.1 | 2 | 162 | ||
NM_005665.5 | NP_005656.4 | 2 | 162 | |||
XM_017002269.1 | XP_016857758.1 | 2 | 160 | |||
XM_017002270.1 | XP_016857759.1 | 2 | 471 | |||
XM_017002271.1 | XP_016857760.1 | 2 | 160 | |||
XM_017002272.1 | XP_016857761.1 | 2 | 160 | |||
XM_017002273.1 | XP_016857762.1 | 2 | 386 | |||
XM_017002274.1 | XP_016857763.1 | 2 | 135 | |||
XM_017002275.1 | XP_016857764.1 | 2 | 123 | |||
XM_017002276.1 | XP_016857765.1 | 2 | 384 | |||
XM_017002277.1 | XP_016857766.1 | 2 | 160 | |||
XM_017002278.1 | XP_016857767.1 | 2 | 160 | |||
XM_017002280.1 | XP_016857769.1 | 2 | 384 | |||
XM_017002281.1 | XP_016857770.1 | 2 | 462 | |||
XM_017002282.1 | XP_016857771.1 | 2 | 160 | |||
XM_017002283.1 | XP_016857772.1 | 2 | 160 | |||
XM_017002284.1 | XP_016857773.1 | 2 | 383 | |||
XM_017002285.1 | XP_016857774.1 | 2 | 383 | |||
GenBank mRNA | AB449875.1 | - | 2 | 152 | ||
AF008915.1 | - | 2 | 163 | |||
BP282704.1 | - | 2 | 162 | |||
CN334472.1 | - | 2 | 359 |