Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009047.5 | NP_033073.1 | 5 | 1220 | ||
GenBank mRNA | AK053133.1 | - | 5 | 1221 | ||
AK084460.1 | - | 5 | 1228 |