Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039150.1 | NP_001034239.1 | 3 | 589 | ||
NM_001039151.1 | NP_001034240.1 | 3 | 589 | |||
NM_001177785.1 | NP_001171256.1 | 3 | 589 | |||
NM_001177786.1 | NP_001171257.1 | 3 | 589 | |||
NM_001177787.1 | NP_001171258.1 | 3 | 589 | |||
NM_009851.2 | NP_033981.2 | 3 | 589 | |||
GenBank mRNA | AJ251594.1 | - | 3 | 376 | ||
AK045226.1 | - | 3 | 435 | |||
AK087472.1 | - | 3 | 383 | |||
AK144125.1 | - | 3 | 392 | |||
AK152099.1 | - | 3 | 385 | |||
AK154411.1 | - | 3 | 380 | |||
AK162720.1 | - | 3 | 385 | |||
AK166320.1 | - | 3 | 374 | |||
BC005676.1 | - | 3 | 368 | |||
BC051388.1 | - | 3 | 419 | 4910789 | ||
CT010324.1 | - | 3 | 270 | |||
J05163.1 | - | 3 | 251 | |||
M27129.1 | - | 3 | 296 | |||
M27130.1 | - | 3 | 302 | |||
M30655.1 | - | 3 | 370 | |||
X66081.1 | - | 3 | 273 | |||
X66082.1 | - | 3 | 273 | |||
X66083.1 | - | 3 | 273 | |||
X66084.1 | - | 3 | 273 |