Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010574.2 | NP_034704.1 | 5 | 1887 | ||
GenBank mRNA | AK148520.1 | - | 5 | 2046 | ||
BC029750.1 | - | 5 | 1887 | 3592499 | ||
BC085291.1 | - | 6 | 1899 | 5347048 |