Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011731.3 | NP_035861.2 | 11 | 1978 | ||
GenBank mRNA | AF075261.1 | - | 11 | 1802 | ||
AK076806.1 | - | 12 | 1709 | |||
AK143893.1 | - | 11 | 1951 | |||
AK144038.1 | - | 11 | 1979 | |||
AK163340.1 | - | 12 | 1867 | |||
AK164706.1 | - | 11 | 1823 | |||
AK164713.1 | - | 9 | 3855 | |||
BC013484.1 | - | 11 | 1889 | 4242168 |