Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cndp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289531.1 | NP_001276460.1 | 10, 11 | 1520 | ||
NM_023149.3 | NP_075638.2 | 11, 12 | 1488 | |||
GenBank mRNA | AB046738.1 | - | 11, 12 | 1429 | ||
AK003779.1 | - | 11, 12 | 1409 | |||
AK131124.1 | - | 10, 11 | 1399 | |||
AK149751.1 | - | 10, 11 | 1444 | |||
AK151013.1 | - | 11, 12 | 1410 | |||
AK152442.1 | - | 10, 11 | 1485 | |||
AK152729.1 | - | 10, 11 | 1458 | |||
AK153040.1 | - | 10, 11 | 1459 | |||
AK159129.1 | - | 10, 11 | 1487 | |||
AK159739.1 | - | 11, 12 | 1415 | |||
AK168029.1 | - | 11, 12 | 1427 | |||
AK168652.1 | - | 11, 12 | 1439 | |||
AK168856.1 | - | 9, 10 | 1177 | |||
AK172034.1 | - | 11, 12 | 1410 | |||
AK185765.1 | - | 2, 3 | 172 | |||
AK208750.1 | - | 1, 2 | 123 | |||
AK214219.1 | - | 2, 3 | 172 | |||
BC005532.1 | - | 11, 12 | 1419 | 3496319 |