Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048057.1 | NP_001041522.1 | 4 | 254 | ||
NM_001048058.1 | NP_001041523.1 | 3 | 180 | |||
NM_023372.2 | NP_075861.1 | 4 | 237 | |||
GenBank mRNA | AB037665.1 | - | 4 | 256 | ||
AK002659.1 | - | 4 | 204 | |||
AK010277.1 | - | 4 | 191 | |||
AK010518.2 | - | 4 | 190 | |||
AK012391.1 | - | 4 | 190 | |||
AK151517.1 | - | 4 | 191 | |||
BC034339.1 | - | 4 | 193 | 3985781 | ||
BC047046.1 | - | 3 | 180 | |||
BC055346.1 | - | 4 | 189 | 5003663 | ||
BC081447.1 | - | 3 | 154 | |||
BC094258.1 | - | 3 | 185 | 1245753 | ||
BC094644.1 | - | 3 | 186 | 3328874 | ||
BU921494.1 | - | 4 | 237 | |||
CJ042718.1 | - | 4 | 220 |