Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkiras1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023526.3 | NP_076015.2 | 4 | 792 | ||
GenBank mRNA | AF225707.1 | - | 4 | 649 | ||
AK010285.1 | - | 5 | 965 | |||
AK028539.1 | - | 4 | 780 | |||
AK148040.1 | - | 4 | 718 | |||
AK167340.1 | - | 4 | 689 | |||
BC061092.1 | - | 4 | 697 | 6589347 |