Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB048364.1 | - | 2, 3 | 313 | ||
AK018634.1 | - | 2, 3 | 140 | |||
AK031379.1 | - | 2, 3 | 376 | |||
AK086508.1 | - | 2, 3 | 306 | |||
BC038274.1 | - | 2, 3 | 383 | 4457335 |