Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005708.3 | NP_005699.1 | 2 | 826 | ||
XM_011521044.2 | XP_011519346.1 | 2 | 151 | |||
XM_017020298.1 | XP_016875787.1 | 2 | 2434 | |||
XM_017020299.1 | XP_016875788.1 | 2 | 1201 | |||
XM_017020300.1 | XP_016875789.1 | 2 | 58 | |||
GenBank mRNA | AF105267.1 | - | 2 | 779 | ||
AF111178.1 | - | 2 | 828 | |||
AK074741.1 | - | 2 | 280 | |||
AK130013.1 | - | 2 | 163 | |||
AK290144.1 | - | 2 | 347 | |||
AK304338.1 | - | 2 | 645 | |||
AY358462.1 | - | 2 | 759 | |||
BC106947.2 | - | 2 | 674 | 40034564 |