Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025513.3 | NP_079789.1 | 2, 3 | 491 | ||
GenBank mRNA | AK006281.1 | - | 2, 3 | 387 | ||
AK012689.1 | - | 2, 3 | 492 | |||
BC023669.1 | - | 2, 3 | 486 | 5012335 | ||
BC054085.1 | - | 2, 3 | 496 | 3371550 |