Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163687.1 | NP_001157159.1 | 8 | 985 | ||
NM_025972.4 | NP_080248.2 | 8 | 991 | |||
GenBank mRNA | AB162194.1 | - | 8 | 957 | ||
AK008776.1 | - | 8 | 993 | |||
AK017141.1 | - | 8 | 968 | |||
AK089243.1 | - | 1 | 233 | |||
AK133485.1 | - | 8 | 976 | |||
AK133658.1 | - | 8 | 992 | |||
AK146191.1 | - | 8 | 979 | |||
BC004572.1 | - | 8 | 961 | 3586255 |