Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026155.3 | NP_080431.1 | 5 | 2247 | ||
GenBank mRNA | AK002818.1 | - | 5 | 2248 | ||
AK012912.1 | - | 1 | 1519 | |||
AK076235.1 | - | 5 | 2280 | |||
AK088555.1 | - | 5 | 2252 | |||
AK150255.1 | - | 5 | 2242 | |||
AK162235.1 | - | 3 | 1951 | |||
BC011111.1 | - | 5 | 2253 | 3965897 |