Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bphl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026512.1 | NP_080788.1 | 3 | 254 | ||
GenBank mRNA | AK002907.1 | - | 3 | 252 | ||
AK017801.1 | - | 3 | 257 | |||
AK078232.1 | - | 3 | 299 | |||
AK152712.1 | - | 3 | 264 | |||
AK170099.1 | - | 3 | 257 | |||
BC023146.1 | - | 3 | 239 | 3990118 | ||
BC025162.1 | - | 3 | 254 | 4921451 |