Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bphl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026512.1 | NP_080788.1 | 7 | 1096 | ||
GenBank mRNA | AK002907.1 | - | 7 | 1094 | ||
AK007606.1 | - | 7 | 1091 | |||
AK008202.1 | - | 7 | 1115 | |||
AK078232.1 | - | 7 | 1141 | |||
AK152712.1 | - | 7 | 1106 | |||
AK170099.1 | - | 7 | 1099 | |||
BC023146.1 | - | 7 | 1081 | 3990118 | ||
BC025162.1 | - | 7 | 1096 | 4921451 | ||
BC029322.1 | - | 5 | 731 |