Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mterf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028832.3 | NP_083108.1 | 3 | 1426 | ||
GenBank mRNA | AA285696.1 | - | 1 | 124 | ||
AK047778.1 | - | 3 | 1402 | |||
BC046619.1 | - | 2 | 1373 | 6494011 |