Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029023.3 | NP_083299.3 | 3 | 368 | ||
GenBank mRNA | AF330052.1 | - | 3 | 324 | ||
AK014680.1 | - | 3 | 369 | |||
AK050181.1 | - | 3 | 306 | |||
AK153225.1 | - | 3 | 356 | |||
AK153485.1 | - | 3 | 313 | |||
AK191644.1 | - | 1 | 63 | |||
AK206268.1 | - | 1 | 63 | |||
BC004847.1 | - | 3 | 358 | 2650587 | ||
BC021399.1 | - | 3 | 323 | 4193674 |