Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rictor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030168.3 | NP_084444.3 | 6 | 466 | ||
GenBank mRNA | AK019537.1 | - | 6 | 464 | ||
AK029196.1 | - | 6 | 468 | |||
AK036149.1 | - | 6 | 467 | |||
AK036492.1 | - | 6 | 467 | |||
AK054119.1 | - | 6 | 456 | |||
AK082586.1 | - | 6 | 467 | |||
AK087858.1 | - | 6 | 468 | |||
AY497009.1 | - | 6 | 444 | |||
AY540053.1 | - | 6 | 464 | |||
BC022906.1 | - | 1 | 12 | |||
BC120904.1 | - | 6 | 464 | 40086995 |