Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mydgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080837.2 | NP_543027.1 | 6 | 499 | ||
GenBank mRNA | AK004074.1 | - | 6 | 499 | ||
AK008289.1 | - | 6 | 485 | |||
AK008854.1 | - | 6 | 480 | |||
AK018016.1 | - | 6 | 482 | |||
AK133947.1 | - | 483 | ||||
AK148083.1 | - | 6 | 499 | |||
AK162834.1 | - | 6 | 487 | |||
AY038184.1 | - | 6 | 495 | |||
BC132127.1 | - | 6 | 520 | 40130404 | ||
BC132129.1 | - | 6 | 520 | 40130406 |