Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005736.3 | NP_005727.1 | 3 | 276 | ||
GenBank mRNA | AK297362.1 | - | 3 | 239 | ||
AK302072.1 | - | 2 | 165 | |||
AK312506.1 | - | 3 | 235 | |||
BC000693.2 | - | 3 | 212 | 3347881 | ||
BC026016.2 | - | 3 | 178 | 3009822 | ||
DC384284.1 | - | 3 | 349 | |||
X82206.1 | - | 3 | 234 | |||
Z14978.1 | - | 3 | 216 |