Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134010.2 | NP_598771.1 | 3 | 248 | ||
GenBank mRNA | AK043454.1 | - | 3 | 243 | ||
AK088069.1 | - | 3 | 249 | |||
AK088303.1 | - | 3 | 225 | |||
AK088625.1 | - | 3 | 218 | |||
AK133938.1 | - | 3 | 208 | |||
AK161665.1 | - | 3 | 236 | |||
BC057591.1 | - | 3 | 215 | 6402677 | ||
BC092261.1 | - | 3 | 225 | 2646396 |