Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018346.1 | - | 7 | 605 | ||
AK041864.1 | - | 7 | 601 | |||
AK082629.1 | - | 7 | 608 | |||
AK158546.1 | - | 7 | 598 | |||
AK169761.1 | - | 7 | 605 | |||
BC016073.1 | - | 7 | 549 | 4482130 |