Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASGRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128602.1 | NP_001122074.1 | 4 | 519 | ||
NM_001306086.1 | NP_001293015.1 | 4 | 519 | |||
NM_005739.3 | NP_005730.2 | 4 | 519 | |||
XM_005254114.2 | XP_005254171.1 | 4 | 508 | |||
XM_011521151.2 | XP_011519453.1 | 4 | 508 | |||
XM_017021860.1 | XP_016877349.1 | 4 | 446 | |||
GenBank mRNA | AF081195.1 | - | 4 | 444 | ||
AF081197.1 | - | 3 | 298 | |||
AF106071.1 | - | 4 | 353 | |||
AK295097.1 | - | 3 | 383 | |||
AK314091.1 | - | 4 | 519 | |||
AK316067.1 | - | 4 | 447 | |||
AY634315.2 | - | 4 | 348 | |||
AY858556.2 | - | 4 | 348 | |||
AY954625.4 | - | 4 | 348 | |||
AY966005.1 | - | 4 | 348 | |||
BC067298.1 | - | 4 | 424 | |||
BC109296.1 | - | 4 | 391 | 40034427 | ||
BC109297.1 | - | 4 | 391 | 40034428 |