Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrgprf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145379.2 | NP_663354.1 | 3 | 1493 | ||
GenBank mRNA | AK157988.1 | - | 4 | 1632 | ||
BC019711.1 | - | 3 | 1493 | 4207906 |
Set Membership: |
G-protein coupled receptor |