Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146328.2 | NP_666440.2 | 2 | 571 | ||
GenBank mRNA | BC132347.1 | - | 1 | 515 | 40130624 |