Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glis1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147221.2 | NP_671754.2 | 4 | 1207 | ||
GenBank mRNA | AF220434.1 | - | 2 | 1061 | ||
AF486579.1 | - | 4 | 1206 | |||
BC066157.1 | - | 2 | 642 | 30468539 | ||
JQ043365.1 | - | 3 | 986 |