Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRE11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005590.3 | NP_005581.2 | 3, 4 | 448 | ||
NM_005591.3 | NP_005582.1 | 3, 4 | 341 | |||
XM_005274008.3 | XP_005274065.1 | 1, 2 | 60 | |||
XM_006718842.3 | XP_006718905.1 | 3, 4 | 448 | |||
XM_011542837.2 | XP_011541139.1 | 3, 4 | 870 | |||
XM_017017772.1 | XP_016873261.1 | 3, 4 | 448 | |||
XM_017017773.1 | XP_016873262.1 | 3, 4 | 341 | |||
GenBank mRNA | AF022778.1 | - | 3, 4 | 322 | ||
AF073362.1 | - | 2, 3 | 152 | |||
AK095388.1 | - | 3, 4 | 450 | |||
AK308318.1 | - | 3, 4 | 339 | |||
BC005241.1 | - | 3, 4 | 301 | |||
BC017823.1 | - | 3, 4 | 384 | |||
BC063458.1 | - | 2, 3 | 249 | 5181691 | ||
BF574168.1 | - | 340 | ||||
BT006730.1 | - | 2, 3 | 152 | |||
KU178199.1 | - | 2, 3 | 152 | |||
KU178200.1 | - | 2, 3 | 152 | |||
U37359.1 | - | 3, 4 | 323 |