Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175238.5 | NP_780447.4 | 2, 3 | 152 | ||
GenBank mRNA | AY428571.2 | - | 2, 3 | 157 | ||
AY585206.1 | - | 1, 2 | 145 | |||
CJ101347.1 | - | 2, 3 | 152 | |||
JX255737.1 | - | 1, 2 | 109 |