Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175438.4 | NP_780647.3 | 2 | 355 | ||
GenBank mRNA | AK044712.1 | - | 2 | 151 | ||
BC024133.1 | - | 1 | 73 | |||
BC039281.1 | - | 2 | 139 | 4975491 | ||
BC056226.1 | - | 2 | 239 | 5064715 | ||
BC060650.1 | - | 2 | 355 | 30354228 | ||
CA494657.1 | - | 2 | 392 |