Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037727.2 | NP_001032816.1 | 10 | 1757 | ||
NM_001286610.1 | NP_001273539.1 | 9 | 1557 | |||
NM_175476.4 | NP_780685.2 | 10 | 1428 | |||
GenBank mRNA | AK037710.1 | - | 10 | 1490 | ||
AK048450.1 | - | 10 | 1388 | |||
AK137444.1 | - | 10 | 1416 | |||
AK137526.1 | - | 10 | 1828 | |||
AK159598.1 | - | 9 | 1558 | |||
AK159941.1 | - | 9 | 1558 | |||
AK172878.1 | - | 8 | 1087 | |||
BC108400.1 | - | 7 | 1038 | |||
BC138751.1 | - | 9 | 1336 | 8861773 | ||
BC138752.1 | - | 9 | 1336 | 8861774 |