Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175482.3 | NP_780691.2 | 7 | 755 | ||
GenBank mRNA | BC066033.1 | - | 7 | 726 | 6835629 | |
BC088733.1 | - | 7 | 677 | 30459522 | ||
CA556001.1 | - | 3 | 168 |