Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199593.1 | NP_001186522.1 | 3 | 541 | ||
NM_177262.4 | NP_796236.2 | 3 | 652 | |||
GenBank mRNA | AK050643.1 | - | 3 | 641 | ||
AK089431.1 | - | 3 | 516 | |||
AK134792.1 | - | 4 | 635 | |||
AK149649.1 | - | 3 | 515 | |||
AK150062.1 | - | 3 | 478 | |||
AK170556.1 | - | 3 | 609 | |||
AK171297.1 | - | 3 | 524 | |||
BC052923.1 | - | 3 | 599 | 6312892 |